LIF019493731 Life Chemicals Inc.

4-cyclohexyl-1,3-thiazol-2-amine

  • MDL number MFCD03473352
  • Cas Number : 7496-55-1
  • Molecular Weight : 182.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart