LIF019493761 Life Chemicals Inc.

5-(Methylamino)nicotinic acid

  • MDL number MFCD04971939
  • Cas Number : 91702-88-4
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart