LIF019493762 Life Chemicals Inc.

5-(dimethylamino)pyridine-3-carboxylic acid

  • MDL number MFCD04971940
  • Cas Number : 30766-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart