LIF019493765 Life Chemicals Inc.

6-Fluoro-2-isopropylquinoline-4-carboxylic acid

  • MDL number MFCD04971943
  • Cas Number : 887206-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart