LIF019493766 Life Chemicals Inc.

9-Fluorobenzo[h]-1,6-naphthyridine-5-carboxylic acid

  • MDL number MFCD04971944
  • Cas Number : 610279-63-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart