LIF019493767 Life Chemicals Inc.

7-Bromopyrido[2,3-b]pyrazine-6-carboxylic acid

  • MDL number MFCD04971945
  • Cas Number : 887206-72-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart