LIF019493773 Life Chemicals Inc.

3-(1-Pyridin-4-yl-3,4-dihydroisoquinolin-2(1H)-yl)propanoic acid

  • MDL number MFCD04971951
  • Cas Number : 610258-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart