LIF019493778 Life Chemicals Inc.

6-Bromo-3H-imidazo[4,5-b]pyridine-5-carboxylic acid

  • MDL number MFCD04971953
  • Cas Number : 889939-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart