LIF021635414 Life Chemicals Inc.

8-Methoxy-2-oxo-2H-chromene-3-carboxylic acid

  • MDL number MFCD01116499
  • Cas Number : 2555-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart