LIF021640777 Life Chemicals Inc.

2-(4-acetylpiperazine-1-carbonyl)benzoic acid

  • MDL number MFCD05862406
  • Cas Number : 890092-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart