LIF021641467 Life Chemicals Inc.

4-(4-Acetylpiperazin-1-yl)benzaldehyde

  • MDL number MFCD05862988
  • Cas Number : 890092-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart