LIF040883113 Life Chemicals Inc.

{[4-(3,4-Dimethoxyphenyl)tetrahydro-2H-pyran-4-yl]methyl}amine hydrochloride

  • MDL number MFCD01654152
  • Cas Number : 61861-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart