LIF040887302 Life Chemicals Inc.

4-Oxo-4-{[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]amino}butanoic acid

  • MDL number MFCD01232130
  • Cas Number : 797806-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart