LIF040887305 Life Chemicals Inc.

6-Bromo-1H-imidazo[4,5-b]pyridin-2-ol

  • MDL number MFCD00763285
  • Cas Number : 148038-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart