LIF040887649 Life Chemicals Inc.

(5-Methyl-2-oxo-1,3-benzoxazol-3(2H)-yl)acetic acid

  • MDL number MFCD04358106
  • Cas Number : 767304-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart