LIF040892870 Life Chemicals Inc.

3-hydroxy-1lambda6-thiolane-1,1-dione

  • MDL number MFCD00154867
  • Cas Number : 13031-76-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart