LIF040892886 Life Chemicals Inc.

3-(methylamino)-1lambda6-thiolane-1,1-dione

  • MDL number MFCD00053546
  • Cas Number : 51070-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart