LIF040892978 Life Chemicals Inc.

5-(pyridin-2-yl)thiophene-2-carboxylic acid

  • MDL number MFCD00052899
  • Cas Number : 119082-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart