LIF040892998 Life Chemicals Inc.

2-bromopyridine-3-carbonitrile

  • MDL number MFCD00234140
  • Cas Number : 20577-26-8
  • Molecular Weight : 183.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart