LIF040893021 Life Chemicals Inc.

4-Imidazo[2,1-b]thiazol-6-yl-phenylamine

  • MDL number MFCD01624149
  • Cas Number : 183668-02-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart