LIF047697272 Life Chemicals Inc.

3-[1-(2-Thienyl)-3,4-dihydroisoquinolin-2(1H)-yl]propanoic acid hydrochloride

  • MDL number MFCD06202334
  • Cas Number : 1052538-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart