LIF049037335 Life Chemicals Inc.

N-Allyl-2-amino-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxamide

  • MDL number MFCD06496412
  • Cas Number : 887202-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart