LIF061765888 Life Chemicals Inc.

(2-Oxo-benzothiazol-3-yl)-acetic acid

  • MDL number MFCD00796503
  • Cas Number : 945-03-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart