LIF062295762 Life Chemicals Inc.

6-amino-1-(3-chloro-4-fluorophenyl)pyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD06809794
  • Cas Number : 887201-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart