LIF062295828 Life Chemicals Inc.

2-Chloro-5-methoxy-benzothiazole

  • MDL number MFCD00160069
  • Cas Number : 3507-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart