LIF064804919 Life Chemicals Inc.

4-Hydroxy-1,6-dimethyl-1H-pyridin-2-one

  • MDL number MFCD00204498
  • Cas Number : 6052-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart