LIF064805091 Life Chemicals Inc.

2,3-Dihydro-1H-benzo[d]imidazo[1,2-a]imidazole

  • MDL number MFCD00805823
  • Cas Number : 24134-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart