LIF064805100 Life Chemicals Inc.

5-Oxo-1-o-tolyl-pyrrolidine-3-carboxylic acid

  • MDL number MFCD02197522
  • Cas Number : 56617-44-8
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart