LIF064805108 Life Chemicals Inc.

2-Amino-benzothiazole-6-sulfonic acid methylamide

  • MDL number MFCD06496312
  • Cas Number : 18101-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart