LIF064805109 Life Chemicals Inc.

2-Amino-benzothiazole-6-sulfonic acid dimethylamide

  • MDL number MFCD06496313
  • Cas Number : 17901-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart