LIF064805125 Life Chemicals Inc.

2-Amino-benzothiazole-6-carboxylic acid ethyl ester

  • MDL number MFCD00102724
  • Cas Number : 50850-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart