LIF068158955 Life Chemicals Inc.

2-Pyridin-4-yl-1H-benzimidazole

  • MDL number MFCD00453948
  • Cas Number : 2208-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart