LIF068161894 Life Chemicals Inc.

4-Amino-5-trifluoromethyl-4H-[1,2,4]triazole-3-thiol

  • MDL number MFCD00101076
  • Cas Number : 24848-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart