LIF068170059 Life Chemicals Inc.

3-Amino-4,6-dimethyl-thieno[2,3-b]pyridine-2-carbonitrile

  • MDL number MFCD00124851
  • Cas Number : 52505-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart