LIF068183219 Life Chemicals Inc.

(biphenyl-4-yloxy)acetic acid

  • MDL number MFCD00179413
  • Cas Number : 13333-86-3
  • Molecular Weight : 228.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart