LIF068190048 Life Chemicals Inc.

ethyl 4,7-dichloro-8-methylquinoline-3-carboxylate

  • MDL number MFCD00173378
  • Cas Number : 58666-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart