LIF068203824 Life Chemicals Inc.

2-Amino-4-methyl-thiazole-5-carboxylic acid dimethylamide

  • MDL number MFCD01459892
  • Cas Number : 21709-40-0
  • Molecular Weight : 185.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart