LIF068205487 Life Chemicals Inc.

2-(4-chlorophenyl)imidazo[1,2-a]pyridin-3-amine

  • MDL number MFCD02078279
  • Cas Number : 34164-94-8
  • Molecular Weight : 243.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart