LIF068212905 Life Chemicals Inc.

1-Isobutyl-1H-benzimidazole-2-carbaldehyde

  • MDL number MFCD03130244
  • Cas Number : 610275-02-0
  • Molecular Weight : 202.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart