LIF068212908 Life Chemicals Inc.

1-(propan-2-yl)-1H-1,3-benzodiazole-2-carbaldehyde

  • MDL number MFCD04373175
  • Cas Number : 339547-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart