LIF068212913 Life Chemicals Inc.

5-Amino-3-mercapto-[1,2,4]triazolo[4,3-a]pyrimidin-7-ol

  • MDL number MFCD01950933
  • Cas Number : 41266-68-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart