LIF068212941 Life Chemicals Inc.

5-Bromo-4,6-dimethyl-nicotinic acid

  • MDL number MFCD07366090
  • Cas Number : 93350-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart