LIF068212944 Life Chemicals Inc.

5-bromo-4,6-dimethylpyridin-3-amine

  • MDL number MFCD07439966
  • Cas Number : 890092-29-2
  • Molecular Weight : 201.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart