LIF068212945 Life Chemicals Inc.

5-Chloro-4,6-dimethylnicotinonitrile

  • MDL number MFCD07439967
  • Cas Number : 890092-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart