LIF068213006 Life Chemicals Inc.

2-(4-Phenyl-4H-[1,2,4]triazol-3-yl)-phenol

  • MDL number MFCD00723849
  • Cas Number : 25222-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart