LIF068213104 Life Chemicals Inc.

(1H-Benzimidazol-2-ylmethyl)formamide

  • MDL number MFCD06643534
  • Cas Number : 879918-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart