LIF069950699 Life Chemicals Inc.

4-Amino-5-(2-methoxy-phenyl)-4H-[1,2,4]triazole-3-thiol

  • MDL number MFCD01912738
  • Cas Number : 61019-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart