LIF069950702 Life Chemicals Inc.

3-(1H-Tetrazol-1-yl)benzoic acid

  • MDL number MFCD00574247
  • Cas Number : 204196-80-5
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart