LIF069953091 Life Chemicals Inc.

2-Chloromethyl-6-ethyl-3H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD01850755
  • Cas Number : 91225-68-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart