LIF069953095 Life Chemicals Inc.

2-methyl-5-oxo-5H-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylic acid

  • MDL number MFCD06254723
  • Cas Number : 32278-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart